Få mere at vide
Propionaldehyde-2,2-d2, TRC
Artikelnummer. 30412836
Skift visning
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30412836 5 mg
30423460 50 mg
30412838 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30412836 Leverandør Toronto Research Chemicals Leverandørnr. TRCP7825035MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 39493-21-5
Molekylær formel C3 D2 H4 O
Molekylvægt (g/mol) 60.09
Synonym Propanal-2,2-d2 (9CI), 2,2-Dideuteropropanal
IUPAC navn 2,2-dideuteriopropanal
SMIL [2H]C([2H])(C)C=O
Tekniske data
Kemisk navn eller materiale Propionaldehyde-2,2-d2
Mængde 5 mg
InChI formel InChI=1S/C3H6O/c1-2-3-4/h3H,2H2,1H3/i2D2
Formel vægt 60.0544