Få mere at vide
Propionaldehyde-2,2,3,3,3-d5, TRC
Artikelnummer. 30413997
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30413997 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30413997 Leverandør Toronto Research Chemicals Leverandørnr. TRCP78250150MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Propionaldehyde-2,2,3,3,3-d5
CAS 198710-93-9
Molekylær formel C3 D5 H O
Mængde 50 mg
Synonym Propanal-2,2,3,3,3-d5 (9CI), 2,2,3,3,3-Pentadeuteriopropanal, Propionaldehyde-d5, Propionaldehyde-2,2,3,3,3-d5, Propanal-2,2,3,3,3-D5
InChI formel InChI=1S/C3H6O/c1-2-3-4/h3H,2H2,1H3/i1D3,2D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C=O
IUPAC navn 2,2,3,3,3-pentadeuteriopropanal
Molekylvægt (g/mol) 63.11
Formel vægt 63.0732