Få mere at vide
Propionaldehyde-2,2,3,3,3-d5, CDN
Artikelnummer. 30319345
Skift visning
Mængde:
0.1 g
0.25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30319345 0.1 g
30316861 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319345 Leverandør CDN Leverandørnr. CDND05590.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Propionaldehyde-2,2,3,3,3-d5
CAS 198710-93-9
Molekylær formel C3 D5 H O
Mængde 0.1 g
Synonym Propanal-2,2,3,3,3-d5 (9 CI), 2,2,3,3,3-Pentadeuteriopropanal, Propionaldehyde-d5, Propionaldehyde-2,2,3,3,3-d5, Propanal-2,2,3,3,3-D5
InChI formel InChI=1 S/C3H6O/c1-2-3-4/h3H,2H2,1H3/i1D3,2D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])C=O
IUPAC navn 2,2,3,3,3-pentadeuteriopropanal
Molekylvægt (g/mol) 63.11
Formel vægt 63.0732 g/mol
Procent renhed 98 atom % D, min 96% Chemical Purity