Learn More
Propiolamide, 98%, BLD Pharm™
Product Code. 31431167
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
10 g
25 g
100 g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31431167 25 g
31431133 100 g
31427572 1 g
31423296 5 g
31415230 250 mg
31426864 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 31431167 Supplier BLD Pharm Supplier No. BD14934625g
 Restricted Item
Forventet forsendelse: 25-11-2026
Specifications
CAS 7341-96-0
Molecular Formula C3H3NO
MDL Number MFCD04035573
Quantity 25 g
SMILES C#CC(N)=O
Molecular Weight (g/mol) 69.06
Percent Purity 98%
Chemical Name or Material Propiolamide