Få mere at vide
Propio-d5-phenone, CDN
Artikelnummer. 30327250
Skift visning
Mængde:
0.5 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30327250 1 g
30319164 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30327250 Leverandør CDN Leverandørnr. CDND58611G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Propiophenone-D5 (ethyl-D5)
CAS 342610-99-5
Molekylær formel C9 D5 H5 O
Mængde 1 g
Synonym 1-Propanone-2,2,3,3,3-d5, 1-phenyl- (9 CI), Propiophenone-d5, Propiophenone-2,2,3,3,3-d5, Propiophenone-D5 (ethyl-D5), 1-Phenyl-1-propanone-2,2,3,3,3-d5
InChI formel InChI=1 S/C9H10O/c1-2-9(10)8-6-4-3-5-7-8/h3-7 H,2H2,1H3/i1D3,2D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])C(=O)c1ccccc1
IUPAC navn 2,2,3,3,3-pentadeuterio-1-phenylpropan-1-one
Molekylvægt (g/mol) 139.206
Formel vægt 139.105 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity