Få mere at vide
Propazine-d14 (di-iso-propyl-d14), CDN
Artikelnummer. 30319921
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30319921 0.05 g
30310681 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319921 Leverandør CDN Leverandørnr. CDND60220.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Propazine D14 (Diisopropyl D14)
CAS 1219802-87-5
Molekylær formel C9 D14 H2 Cl N5
Mængde 0.05 g
Synonym Propazine D14 (Diisopropyl D14)
InChI formel InChI=1 S/C9H16ClN5/c1-5(2)11-8-13-7(10)14-9(15-8)12-6(3)4/h5-6 H,1-4H3,(H2,11,12,13,14,15)/i1D3,2D3,3D3,4D3,5 D,6 D
SMIL [2 H]C([2 H])([2 H])C([2 H])(Nc1nc(Cl)nc(NC([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H])n1)C([2 H])([2 H])[2 H]
IUPAC navn 6-chloro-2-N,4-N-bis(1,1,1,2,3,3,3-heptadeuteriopropan-2-yl)-1,3,5-triazine-2,4-diamine
Molekylvægt (g/mol) 243.796
Formel vægt 243.197 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity