missing translation for 'onlineSavingsMsg'
Få mere at vide

Propargylsulfid, TRC

Artikelnummer. 30419726
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30419726 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30419726 Leverandør Toronto Research Chemicals Leverandørnr. TRCP839618100MG
 Begrænset vare
Forventet forsendelse: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Højrenheds organiske molekyler og analytiske standarder, strategisk leveret verden over for at muliggøre innovation og kommerciel succes.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Propargyl Sulfide
CAS 13702-09-5
Molekylær formel C6H6S
Mængde 100 mg
InChI formel InChI=1S/C6H6S/c1-3-5-7-6-4-2/h1-2H,5-6H2
SMIL C#CCSCC#C
IUPAC navn 3-prop-2-ynylsulfanylprop-1-yne
Molekylvægt (g/mol) 110.18
Formel vægt 110.019
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.