Beskrivelse
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
- Expert Technical Support
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Propaquizafop |
| Emballage | Glass Bottle |
| CAS | 111479-05-1 |
| Smeltepunkt | 62°C to 64°C |
| Synonym | 2-isopropylideneaminooxyethyl (R)-2-[4-(6-chloroquinoxalin-2-yloxy)phenoxy]propionate |
| Industry Type | Environmental Analysis |
| Mængde | 100 mg |
| Grad | Analytical Standard |