missing translation for 'onlineSavingsMsg'
Få mere at vide

Propantheline Bromide, TRC

Artikelnummer. 30409193
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30409193 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30409193 Leverandør Toronto Research Chemicals Leverandørnr. TRCP760900100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Propantheline Bromide
CAS 50-34-0
Molekylær formel C23 H30 N O3 . Br
Mængde 100 mg
Synonym Propantheline bromide, N-Methyl-N,N-bis(1-methylethyl)-2-[(9H-xanthen-9-ylcarbonyl)oxy]ethanaminium bromide, (2-Hydroxyethyl)diisopropylmethylammonium bromide xanthene-9-carboxylate (6CI), 2-Propanaminium','N-methyl-N-(1-methylethyl)-N-[2-[(9H-xanthen-9-ylcarbonyl)oxy]ethyl]-','bromide (9CI), Ammonium','(2-hydroxyethyl)diisopropylmethyl-','bromide','xanthene-9-carboxylate (8CI), Corrigast, Ercorax, Ercotina, Ketaman, Kivatin, Neo-Metantyl, Neopepulsan, Pantas, Pantheline, Pervagal, Pro-Banthine, Pro-Gastron, Prodixamon, Propantel, Propantheline bromide, SC 3171, Xanthene-9-carboxylic acid','ester with (2-hydroxyethyl)diisopropylmethylammonium bromide, β-Diisopropylaminoethyl xanthene-9-carboxylate methobromide
InChI formel InChI=1S/C23H30NO3.BrH/c1-16(2)24(5,17(3)4)14-15-26-23(25)22-18-10-6-8-12-20(18)27-21-13-9-7-11-19(21)22;/h6-13,16-17,22H,14-15H2,1-5H3;1H/q+1;/p-1
SMIL [Br-].CC(C)[N+](C)(CCOC(=O)C1c2ccccc2Oc3ccccc13)C(C)C
IUPAC navn methyl-di(propan-2-yl)-[2-(9H-xanthene-9-carbonyloxy)ethyl]azanium;bromide
Molekylvægt (g/mol) 448.39
Formel vægt 447.1409
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.