Få mere at vide
Propanil-d5, TRC
Artikelnummer. 30421729
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30421729 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30421729 Leverandør Toronto Research Chemicals Leverandørnr. TRCP7608425MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Propanil-d5
CAS 1398065-82-1
Molekylær formel C9 D5 H4 Cl2 N O
Mængde 5 mg
Synonym Propanil-d5, Propanil-D5 (propanoyl-D5), 3',4'-Dichloropropionanilide-2,2,3,3,3-d5, N-(3,4-Dichlorophenyl)propanamide-2,2,3,3,3-d5, N-(3,4-Dichlorophenyl)propionamide-2,2,3,3,3-d5
InChI formel InChI=1S/C9H9Cl2NO/c1-2-9(13)12-6-3-4-7(10)8(11)5-6/h3-5H,2H2,1H3,(H,12,13)/i1D3,2D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C(=O)Nc1ccc(Cl)c(Cl)c1
IUPAC navn 2,2,3,3,3-pentadeuterio-N-(3,4-dichlorophenyl)propanamide
Molekylvægt (g/mol) 223.111
Formel vægt 222.038