Få mere at vide
Propamocarb-N-oxide, TRC
Artikelnummer. 30135909
Skift visning
Mængde:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135909 500 mg
17980985 250 mg
17941695 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30135909 Leverandør Toronto Research Chemicals Leverandørnr. TRCP758465500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 743449-09-4
Molekylær formel C9 H20 N2 O3
Molekylvægt (g/mol) 204.2667
Synonym Propamocarb-N-oxide, Propyl N-[3-(dimethyloxidoamino)propyl]carbamate (ACI), Carbamic acid, N-[3-(dimethyloxidoamino)propyl]-, propyl ester (ACI), Carbamic acid, [3-(dimethyloxidoamino)propyl]-, propyl ester (9CI), Propyl N-[3-(dimethyloxidoamino)propyl]carbamate (ACI), AE B155306
IUPAC navn N,N-dimethyl-3-(propoxycarbonylamino)propan-1-amine oxide
SMIL CCCOC(=O)NCCC[N+](C)(C)[O-]
Tekniske data
Kemisk navn eller materiale Propamocarb-N-oxide
Mængde 500 mg
InChI formel InChI=1S/C9H20N2O3/c1-4-8-14-9(12)10-6-5-7-11(2,3)13/h4-8H2,1-3H3,(H,10,12)
Formel vægt 204.1474