Få mere at vide
Propamocarb-d7, TRC
Artikelnummer. 30423819
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30423819 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30423819 Leverandør Toronto Research Chemicals Leverandørnr. TRCP7584632.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Propamocarb free base D7 (O-propyl D7)
CAS 1398065-89-8
Molekylær formel C9 2H7 H13 N2 O2
Mængde 2.5 mg
Synonym Propamocarb free base D7 (O-propyl D7), Propamocarb free base D7
InChI formel InChI=1S/C9H20N2O2/c1-4-8-13-9(12)10-6-5-7-11(2)3/h4-8H2,1-3H3,(H,10,12)/i1D3,4D2,8D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])OC(=O)NCCCN(C)C
IUPAC navn 1,1,2,2,3,3,3-heptadeuteriopropyl N-[3-(dimethylamino)propyl]carbamate
Molekylvægt (g/mol) 195.31
Formel vægt 195.1964