Få mere at vide
Propafenone Hydrochloride, TRC
Artikelnummer. 30003624
Skift visning
Mængde:
10 mg
25 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30003624 250 mg
17931895 25 mg
17922235 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30003624 Leverandør Toronto Research Chemicals Leverandørnr. TRCP757500250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 34183-22-7
Molekylær formel C21 H27 N O3 . Cl H
Molekylvægt (g/mol) 377.9
Synonym Propafenone Hydrochloride, 1-Propanone, 1-[2-[2-hydroxy-3-(propylamino)propoxy]phenyl]-3-phenyl-, hydrochloride (1:1), 1-Propanone, 1-[2-[2-hydroxy-3-(propylamino)propoxy]phenyl]-3-phenyl-, hydrochloride (9CI), Propiophenone, 2'-[2-hydroxy-3-(propylamino)propoxy]-3-phenyl-, hydrochloride (8CI), Arythmol, Pronon, Propafenone hydrochloride, Rythmol, Rytmonorm
IUPAC navn 1-[2-[2-hydroxy-3-(propylamino)propoxy]phenyl]-3-phenylpropan-1-one;hydrochloride
SMIL Cl.CCCNCC(O)COc1ccccc1C(=O)CCc2ccccc2
Tekniske data
Kemisk navn eller materiale Propafenone Hydrochloride
Mængde 250 mg
InChI formel InChI=1S/C21H27NO3.ClH/c1-2-14-22-15-18(23)16-25-21-11-7-6-10-19(21)20(24)13-12-17-8-4-3-5-9-17;/h3-11,18,22-23H,2,12-16H2,1H3;1H
Formel vægt 377.1758