Få mere at vide
Propafenone Hydrochloride, LoGiCal
Artikelnummer. 30459293
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30459293 10 mg
1 options
Artikelnummer. 30459293 Leverandør LoGiCal Leverandørnr. LGCFOR0405.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Propafenone Hydrochloride
CAS 34183-22-7
Analyt- eller komponentnavne Propafenone Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C21 H27 N O3 . Cl H
Certificeringer/compliance ISO 9001
Synonym Propafenone Hydrochloride, 1-Propanone, 1-[2-[2-hydroxy-3-(propylamino)propoxy]phenyl]-3-phenyl-, hydrochloride (1:1), 1-Propanone, 1-[2-[2-hydroxy-3-(propylamino)propoxy]phenyl]-3-phenyl-, hydrochloride (9CI), Propiophenone, 2'-[2-hydroxy-3-(propylamino)propoxy]-3-phenyl-, hydrochloride (8CI), Arythmol, Pronon, Propafenone hydrochloride, Rythmol, Rytmonorm
InChI formel InChI=1S/C21H27NO3.ClH/c1-2-14-22-15-18(23)16-25-21-11-7-6-10-19(21)20(24)13-12-17-8-4-3-5-9-17;/h3-11,18,22-23H,2,12-16H2,1H3;1H
Løsningstype Neat
SMIL Cl.CCCNCC(O)COc1ccccc1C(=O)CCc2ccccc2
IUPAC navn 1-[2-[2-hydroxy-3-(propylamino)propoxy]phenyl]-3-phenylpropan-1-one;hydrochloride
Industry Type Clinical Testing
Molekylvægt (g/mol) 377.9
Mængde 10 mg
Formel vægt 377.1758 g/mol