Få mere at vide
Propafenone EP Impurity A, Mikromol
Artikelnummer. 30463209
Skift visning
Mængde:
25 mg
4 x 25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30463209 4 x 25 mg
30052410 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30463209 Leverandør Mikromol Leverandørnr. MM0405.0200254AMP
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Propafenone EP Impurity A, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-(2-Hydroxyphenyl)-3-phenylpropan-1-one
CAS 3516-95-8
Analyt- eller komponentnavne 1-(2-Hydroxyphenyl)-3-phenylpropan-1-one
Forsendelsestilstand Room Temperature
Molekylær formel C15 H14 O2
Certificeringer/compliance ISO 9001
Synonym Propafenone Hydrochloride Imp. A (EP), Propafenone Imp. A (EP), 2'-Hydroxy-3-phenylpropiophenone, Propafenone Hydrochloride Impurity A, Propafenone Impurity A
InChI formel InChI=1S/C15H14O2/c16-14-9-5-4-8-13(14)15(17)11-10-12-6-2-1-3-7-12/h1-9,16H,10-11H2
Løsningstype Neat
SMIL Oc1ccccc1C(=O)CCc2ccccc2
IUPAC navn 1-(2-hydroxyphenyl)-3-phenylpropan-1-one
Molekylvægt (g/mol) 226.27
Mængde 4 x 25 mg
Formel vægt 226.0994 g/mol