Få mere at vide
Propafenone Dimer Impurity-d10, TRC
Artikelnummer. 30419562
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30419562 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30419562 Leverandør Toronto Research Chemicals Leverandørnr. TRCP7575221MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Propafenone Dimer Impurity-d10
CAS 1346602-27-4
Molekylær formel C39H35D10NO6
Mængde 1 mg
Synonym 1,1’-[Propyliminobis[[2-hydroxy(propane-d5)-1,3-diyl]oxy-2,1-phenylene]]bis(3-phenylpropan-1-one), Propafenone Impurity G-d10
InChI formel InChI=1S/C39H45NO6/c1-2-25-40(26-32(41)28-45-38-19-11-9-17-34(38)36(43)23-21-30-13-5-3-6-14-30)27-33(42)29-46-39-20-12-10-18-35(39)37(44)24-22-31-15-7-4-8-16-31/h3-20,32-33,41-42H,2,21-29H2,1H3/i26D2,27D2,28D2,29D2,32D,33D
SMIL [2H]C([2H])(Oc1ccccc1C(=O)CCc2ccccc2)C([2H])(O)C([2H])([2H])N(CCC)C([2H])([2H])C([2H])(O)C([2H])([2H])Oc3ccccc3C(=O)CCc4ccccc4
IUPAC navn 1-[2-[1,1,2,3,3-pentadeuterio-2-hydroxy-3-[[1,1,2,3,3-pentadeuterio-2-hydroxy-3-[2-(3-phenylpropanoyl)phenoxy]propyl]-propylamino]propoxy]phenyl]-3-phenylpropan-1-one
Molekylvægt (g/mol) 633.64
Formel vægt 633.3875