Få mere at vide
Propafenone-d5 Hydrochloride, TRC
Artikelnummer. 30410514
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30410514 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30410514 Leverandør Toronto Research Chemicals Leverandørnr. TRCP7575041MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Propafenone 2,2,3,3,3-propyl-d5 Hydrochloride
CAS 1398066-02-8
Molekylær formel C21 D5 H22 N O3 . Cl H
Mængde 1 mg
Synonym 1-[2-[2-Hydroxy-3-(propylamino)propoxy]phenyl]-3-phenyl-d5-1-propanone Hydrochloride, 1-[2-[2-Hydroxy-3-(propyl-2,2,3,3,3-d5-amino)propoxy]phenyl]-3-phenyl-1-propanone hydrochloride, 1-[2-[2-Hydroxy-3-(pentadeuteriopropylamino)propoxy]phenyl]-3-phenylpropan-1-one hydrochloride, 2'-[2-Hydroxy-3-(propylamino)propoxy]-3-phenyl-d5-propiophenone Hydrochloride, Arythmol-d5, Pronon-d5, Rythmol-d5, Rytmonorm-d5, Propafenone-D5 (propyl-2,2,3,3,3-D5) hydrochloride, Propafenone-d5 hydrochloride
InChI formel InChI=1S/C21H27NO3.ClH/c1-2-14-22-15-18(23)16-25-21-11-7-6-10-19(21)20(24)13-12-17-8-4-3-5-9-17;/h3-11,18,22-23H,2,12-16H2,1H3;1H/i1D3,2D2;
SMIL Cl.[2H]C([2H])([2H])C([2H])([2H])CNCC(O)COc1ccccc1C(=O)CCc2ccccc2
IUPAC navn 1-[2-[2-hydroxy-3-(2,2,3,3,3-pentadeuteriopropylamino)propoxy]phenyl]-3-phenylpropan-1-one;hydrochloride
Molekylvægt (g/mol) 382.94
Formel vægt 382.2072