missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Propacetamol Hydrochloride, TRC
Specifications
Specifications
| Kemisk navn eller materiale | Propacetamol Hydrochloride |
| CAS | 66532-86-3 |
| Molekylær formel | C14 H20 N2 O3 . Cl H |
| Mængde | 500 mg |
| Synonym | Propacetamol hydrochloride, 4-(Acetylamino)phenyl (diethylamino)acetate hydrochloride, Glycine','N,N-diethyl-','4-(acetylamino)phenyl ester','hydrochloride (1:1), Glycine','N,N-diethyl-','4-(acetylamino)phenyl ester','monohydrochloride (9CI), Pro-Dafalgan, Pro-efferalgan, UP 34101 |
| InChI formel | InChI=1S/C14H20N2O3.ClH/c1-4-16(5-2)10-14(18)19-13-8-6-12(7-9-13)15-11(3)17;/h6-9H,4-5,10H2,1-3H3,(H,15,17);1H |
| SMIL | Cl.CCN(CC)CC(=O)Oc1ccc(NC(=O)C)cc1 |
| IUPAC navn | (4-acetamidophenyl) 2-(diethylamino)acetate;hydrochloride |
| Molekylvægt (g/mol) | 300.78 |
| Formel vægt | 300.1241 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?