Få mere at vide
Prometon-d3 (methoxy-d3), TRC
Artikelnummer. 30425519
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30425519 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30425519 Leverandør Toronto Research Chemicals Leverandørnr. TRCP75703610MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Prometon methoxy-d3
CAS 1219803-43-6
Molekylær formel C10 2H3 H16 N5 O
Mængde 10 mg
Synonym Prometon D3 (methyl D3)
InChI formel InChI=1S/C10H19N5O/c1-6(2)11-8-13-9(12-7(3)4)15-10(14-8)16-5/h6-7H,1-5H3,(H2,11,12,13,14,15)/i5D3
SMIL [2H]C([2H])([2H])Oc1nc(NC(C)C)nc(NC(C)C)n1
IUPAC navn 2-N,4-N-di(propan-2-yl)-6-(trideuteriomethoxy)-1,3,5-triazine-2,4-diamine
Molekylvægt (g/mol) 228.31
Formel vægt 228.1778