Få mere at vide
Promethazine Sulfoxide-d6, TRC
Artikelnummer. 30422683
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30422683 1 mg
30409276 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30422683 Leverandør Toronto Research Chemicals Leverandørnr. TRCP7570121MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1189892-93-0
Molekylær formel C17H14D6N2OS
Molekylvægt (g/mol) 306.46
Synonym N,N,-Di(methyl-d6),α-methyl-10H-phenothiazine-10-ethanamine 5-Oxide, N-[2-(Dimethyl-d6-amino)propyl]phenothiazine S-Oxide, Promethazine 5-Oxide-d6, Promethazine 5-Sulfoxide-d6, Romergan Sulfoxide-d6
SMIL CC(N(C([2H])([2H])[2H])C([2H])([2H])[2H])CN1C2=C(C=CC=C2)S(C3=CC=CC=C31)=O
Tekniske data
Kemisk navn eller materiale Promethazine Sulfoxide-d6
Renhedsgrad noter HPLC
Mængde 1 mg
InChI formel InChI=1S/C17H20N2OS/c1-13(18(2)3)12-19-14-8-4-6-10-16(14)21(20)17-11-7-5-9-15(17)19/h4-11,13H,12H2,1-3H3/i2D3,3D3
Formel vægt 306.1673
Procent renhed >95%