Få mere at vide
Promethazine Sulfone Hydrochloride, Mikromol™
Beskrivelse
Promethazine Sulfone Hydrochloride, Mikromol
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Promethazine Sulfone Hydrochloride |
| CAS | 15374-15-9 |
| Analyt- eller komponentnavne | Promethazine Sulfone Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C17 H20 N2 O2 S . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | N,N,alpha-Trimethyl-10H-phenothiazine-10-ethanamine 5,5-Dioxide Hydrochloride (1:1), 10H-Phenothiazine-10-ethanamine, N,N,alpha-trimethyl-, 5,5-dioxide, monohydrochloride (9CI), Phenothiazine, 10-(2-dimethylaminopropyl)-, dioxide, hydrochloride (6CI), Phenothiazine, 10-[2-(dimethylamino)propyl]-, 5,5-dioxide, monohydrochloride (8CI), Dioxopromethazine hydrochloride, N-(2-Dimethylaminopropyl)phenothiazine S,S-dioxide hydrochloride |
| InChI formel | InChI=1S/C17H20N2O2S.ClH/c1-13(18(2)3)12-19-14-8-4-6-10-16(14)22(20,21)17-11-7-5-9-15(17)19;/h4-11,13H,12H2,1-3H3;1H |
| Løsningstype | Neat |
| SMIL | Cl.CC(CN1c2ccccc2S(=O)(=O)c3ccccc13)N(C)C |
| Vis mere |