missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Promethazine Hydrochloride, TRC
Specifications
Specifications
| Kemisk navn eller materiale | Promethazine Hydrochloride |
| CAS | 58-33-3 |
| Molekylær formel | C17 H20 N2 S . Cl H |
| Mængde | 50 mg |
| Synonym | Promethazine Hydrochloride, 10H-Phenothiazine-10-ethanamine','N,N,α-trimethyl-','hydrochloride (1:1), 10H-Phenothiazine-10-ethanamine','N,N,α-trimethyl-','monohydrochloride (9CI), Phenothiazine','10-[2-(dimethylamino)propyl]-','monohydrochloride (8CI), 10-[2-(Dimethylamino)propyl]phenothiazine hydrochloride, Anergan 25, Anergan 50, Atosil, Diprasine, Diprazin, Dorme, Duplamin, Fellozine, Fenazil, Fenergan, Genphen, Hiberna, Himazin, Kinetosin, Lergigan, Mepergan, NSC 231688, Phanergan D, Phena, Phencen, Phenergan, Phenergan VC, Pipolfen, Pipolphen, Plletia, Promasun, Promethazine chloride, Promethazine hydrochloride, Promethazine monohydrochloride, Promethazinium chloride, Prorex, Prothazine, Provigan, Pyrethia, Remsed, Romergan, Romergan hydrochloride |
| InChI formel | InChI=1S/C17H20N2S.ClH/c1-13(18(2)3)12-19-14-8-4-6-10-16(14)20-17-11-7-5-9-15(17)19;/h4-11,13H,12H2,1-3H3;1H |
| SMIL | Cl.CC(CN1c2ccccc2Sc3ccccc13)N(C)C |
| IUPAC navn | N,N-dimethyl-1-phenothiazin-10-ylpropan-2-amine;hydrochloride |
| Molekylvægt (g/mol) | 320.88 |
| Formel vægt | 320.1114 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?