Få mere at vide
Promethazine-d6 Hydrochloride, TRC
Artikelnummer. 30422805
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30422805 1 mg
30428373 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30422805 Leverandør Toronto Research Chemicals Leverandørnr. TRCP7570021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1189947-02-1
Molekylær formel C17 2H6 H14 N2 S . Cl H
Molekylvægt (g/mol) 326.92
Synonym 1-Phenothiazin-10-yl-N,N-bis(trideuteriomethyl)propan-2-amine hydrochloride, Promethazine D6 (N,N-dimethyl D6) hydrochloride
IUPAC navn 1-phenothiazin-10-yl-N,N-bis(trideuteriomethyl)propan-2-amine;hydrochloride
SMIL Cl.[2H]C([2H])([2H])N(C(C)CN1c2ccccc2Sc3ccccc13)C([2H])([2H])[2H]
Tekniske data
Kemisk navn eller materiale Promethazine N,N-dimethyl-d6 hydrochloride
Renhedsgrad noter HPLC
Mængde 1 mg
InChI formel InChI=1S/C17H20N2S.ClH/c1-13(18(2)3)12-19-14-8-4-6-10-16(14)20-17-11-7-5-9-15(17)19;/h4-11,13H,12H2,1-3H3;1H/i2D3,3D3;
Formel vægt 326.1491
Procent renhed >95%