Få mere at vide
Promazine Sulphoxide, LoGiCal
Artikelnummer. 30460779
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30460779 10 mg
1 options
Artikelnummer. 30460779 Leverandør LoGiCal Leverandørnr. LGCFOR0290.01
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Promazine Sulphoxide
CAS 146-21-4
Analyt- eller komponentnavne Promazine Sulphoxide
Forsendelsestilstand Room Temperature
Molekylær formel C17 H20 N2 O S
Certificeringer/compliance ISO 9001
Synonym 3-(10H-Phenothiazin-10-yl)-N,N-dimethylpropan-1-amine S-Oxide (Promazine Sulphoxide), Promazine Hydrochloride Imp. A (EP), Promazine Imp. A (EP), 3-(10H-Phenothiazin-10-yl)-N,N-dimethylpropan-1-amine S-Oxide, Promazine Sulphoxide, Promazine Hydrochloride Impurity A, Promazine Impurity A
InChI formel InChI=1S/C17H20N2OS/c1-18(2)12-7-13-19-14-8-3-5-10-16(14)21(20)17-11-6-4-9-15(17)19/h3-6,8-11H,7,12-13H2,1-2H3
Løsningstype Neat
SMIL CN(C)CCCN1c2ccccc2S(=O)c3ccccc13
IUPAC navn N,N-dimethyl-3-(5-oxophenothiazin-10-yl)propan-1-amine
Industry Type Clinical Testing
Molekylvægt (g/mol) 300.42
Mængde 10 mg
Formel vægt 300.1296 g/mol