Få mere at vide
Promazine Hydrochloride, Mikromol™
Beskrivelse
Promazine Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Promazine hydrochloride |
| CAS | 53-60-1 |
| Analyt- eller komponentnavne | Promazine hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C17 H20 N2 S . Cl H |
| Certificeringer/compliance | ISO 17034 |
| Synonym | Chlorpromazine Hydrochloride Imp. C (EP), Chlorpromazine Imp. C (EP), 3-(10H-Phenothiazin-10-yl)-N,N-dimethylpropan-1-amine Hydrochloride, Promazine Hydrochloride, Chlorpromazine Hydrochloride Imp. C (EP) as Hydrochloride, 10H-Phenothiazine-10-propanamine, N,N-dimethyl-, hydrochloride (1:1), 10H-Phenothiazine-10-propanamine, N,N-dimethyl-, monohydrochloride (9CI), Phenothiazine, 10-[3-(dimethylamino)propyl]-, monohydrochloride (8CI), 10-(gamma-Dimethylamino-n-propyl)phenothiazine hydrochloride, Esparinal, Liranol, Prolactyl, Promazine monohydrochloride, Promwill, Propazin, Propazin (pharmaceutical), Propazine, Prozine, Sparine, Talofen, Chlorpromazine Hydrochloride Impurity C as Hydrochloride, Chlorpromazine Impurity C as Hydrochloride |
| InChI formel | InChI=1S/C17H20N2S.ClH/c1-18(2)12-7-13-19-14-8-3-5-10-16(14)20-17-11-6-4-9-15(17)19;/h3-6,8-11H,7,12-13H2,1-2H3;1H |
| SMIL | Cl.CN(C)CCCN1c2ccccc2Sc3ccccc13 |
| IUPAC navn | N,N-dimethyl-3-phenothiazin-10-ylpropan-1-amine;hydrochloride |
| Vis mere |