Få mere at vide
Promazine-d6 Hydrochloride, TRC
Artikelnummer. 30428367
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30428367 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30428367 Leverandør Toronto Research Chemicals Leverandørnr. TRCP7563021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Promazine-d6 Hydrochloride
CAS 1246819-33-9
Molekylær formel C17H15D6ClN2S
Mængde 1 mg
Synonym N,N-(Dimethyl-d6)-10H-phenothiazine-10-propanamine Hydrochloride, 10-[γ-(Dimethyl-d6)amino-n-propyl]phenothiazine Hydrochloride, Esparinal-d6, Liranol-d6, Prolactyl-d6, Promwill-d6, Sparine-d6, Talofen-d6
InChI formel InChI=1S/C17H20N2S.ClH/c1-18(2)12-7-13-19-14-8-3-5-10-16(14)20-17-11-6-4-9-15(17)19;/h3-6,8-11H,7,12-13H2,1-2H3;1H/i1D3,2D3;
SMIL [2H]C([2H])([2H])N(C([2H])([2H])[2H])CCCN1C2=CC=CC=C2SC3=C1C=CC=C3.Cl
IUPAC navn 3-phenothiazin-10-yl-N,N-bis(trideuteriomethyl)propan-1-amine;hydrochloride
Molekylvægt (g/mol) 326.92
Formel vægt 326.1491