Learn More
Invitrogen™ Prolactin Human ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EHIAPRLX5
Additional Details : Weight : 0.33820kg
Includes: Pre-coated plate(s), standard, peroxidase conjugated detector antibody, assay buffer, wash buffer, chromogen, stop solution, plate covers and lot-specific technical data sheet.
Description
The Human Prolactin ELISA quantitates human prolactin in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu Prolactin. Principle of the method The Human Prolactin solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Prolactin (PRL) or Luteotropic hormone (LTH) is a peptide hormone discovered by Dr. Henry Friesen, primarily associated with lactation. In breastfeeding, the act of an infant sucking the nipple stimulates the production of prolactin, which fills the breast with milk via a process called lactogenesis, in preparation for the next feed. Oxytocin, another hormone, is also released, which triggers milk let-down.Specifications
P01236 | |
11.7 pg/mL | |
Multiplex assay on the Luminex Platform | |
Fluorescence | |
Peptide hormone | |
5 x 96 assays | |
ELISA, Protein Assays, Aktivity Assays, Protein Biology, Reproductive Biology, Inflammation, Oxidative Stress, Metabolism, Cell Signaling, Epigenetics, Hematology, DNA Damage, Cancer Biology, Immunology, Neuroscience, Neurobiology, Steroid Hormone Detecti | |
5617 | |
10.8% | |
Standard, Chromogen, Pre-coated 96 well plates, Stop Solution, Assay Diluent Concentrate, HRP-Conjugated Detection Antibody, Wash Buffer, Adhesive Plate Covers | |
5 x 96 Tests | |
Reproductive Biology | |
-20°C | |
1 hr. 20 min. |
15.6 pg/mL to 1000 pg/mL | |
Human Prolactin ELISA Kit | |
HRP | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
Human | |
Colorimetric Microplate Reader | |
PRL | |
7.6% | |
HRP | |
RUO | |
Plasma, 12.5 μ;L; Serum, 12.5 μ;L; Supernatant, 50 μ;L | |
Human | |
2 hr. 30 min. |