Få mere at vide
Tocris Bioscience™ ProK
GREENER_CHOICE
Artikelnummer. 18758496
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18758496 10 mg
18707696 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18758496 Leverandør Tocris Bioscience™ Leverandørnr. 7361/10
Unnatural amino acid, propargyl lysine derivative
ProK is a lysine-based unnatural amino acid (UAA) used for genetic code expansion. It enables bioconjugation of fluorescent probes in E. coli or multicellular organisms.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N6-[(2-Propyn-1-yloxy)carbonyl]-L-lysine hydrochloride
CAS 1428330-91-9
Mængde 10 mg
Mål Unnatural Amino Acids
Molekylær formel C10H16N2O4.HCl
Renhed 0.95