Få mere at vide
Proguanil EP Impurity D (as Hydrochloride), Mikromol™
Artikelnummer. 30464051
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30464051 4 x 25 mg
30050837 25 mg
17973976 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30464051 Leverandør Mikromol Leverandørnr. MM0840.0500254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Proguanil EP Impurity D (as Hydrochloride), Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,5-bis(1-methylethyl)biguanide hydrochloride
CAS 35708-82-8
Analyt- eller komponentnavne 1,5-bis(1-methylethyl)biguanide hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C8 H19 N5 . Cl H
Certificeringer/compliance ISO 17025
Synonym 1,5-Bis(1-methylethyl)biguanide Hydrochloride, Imidodicarbonimidic diamide, N,N'-bis(1-methylethyl)-, hydrochloride (1:1), Imidodicarbonimidic diamide, N,N'-bis(1-methylethyl)-, monohydrochloride (9CI), GS 18667, Proguanil Hydrochloride Imp. D (EP), Proguanil Imp. D (EP), Proguanil USP Related Compound D, Proguanil USP RC D, 5629W95, Ph Eur Proguanil Impurity D, USP Proguanil Related Compound D, Proguanil Related Compound D as Hydrochloride, Proguanil Hydrochloride Impurity D as Hydrochloride, Proguanil Impurity D as Hydrochloride
InChI formel InChI=1S/C8H19N5.ClH/c1-5(2)11-7(9)13-8(10)12-6(3)4;/h5-6H,1-4H3,(H5,9,10,11,12,13);1H
Løsningstype Neat
SMIL Cl.CC(C)NC(=N)NC(=N)NC(C)C
IUPAC navn 1-propan-2-yl-3-(N-propan-2-ylcarbamimidoyl)guanidine;hydrochloride
Molekylvægt (g/mol) 221.73
Mængde 4 x 25 mg
Formel vægt 221.1407 g/mol