Få mere at vide
Proguanil EP Impurity D (as Hydrochloride), Mikromol™
Beskrivelse
Proguanil EP Impurity D (as Hydrochloride)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 1,5-bis(1-methylethyl)biguanide hydrochloride |
| CAS | 35708-82-8 |
| Analyt- eller komponentnavne | 1,5-bis(1-methylethyl)biguanide hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C8 H19 N5 . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | 1,5-Bis(1-methylethyl)biguanide Hydrochloride, Imidodicarbonimidic diamide, N,N'-bis(1-methylethyl)-, hydrochloride (1:1), Imidodicarbonimidic diamide, N,N'-bis(1-methylethyl)-, monohydrochloride (9CI), GS 18667, Proguanil Hydrochloride Imp. D (EP), Proguanil Imp. D (EP), Proguanil USP Related Compound D, Proguanil USP RC D, 5629W95, Ph Eur Proguanil Impurity D, USP Proguanil Related Compound D, Proguanil Related Compound D as Hydrochloride, Proguanil Hydrochloride Impurity D as Hydrochloride, Proguanil Impurity D as Hydrochloride |
| InChI formel | InChI=1S/C8H19N5.ClH/c1-5(2)11-7(9)13-8(10)12-6(3)4;/h5-6H,1-4H3,(H5,9,10,11,12,13);1H |
| SMIL | Cl.CC(C)NC(=N)NC(=N)NC(C)C |
| IUPAC navn | 1-propan-2-yl-3-(N-propan-2-ylcarbamimidoyl)guanidine;hydrochloride |
| Vis mere |