missing translation for 'onlineSavingsMsg'
Få mere at vide

Progesterone-d9, TRC

Artikelnummer. 30419099
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30419099 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30419099 Leverandør Toronto Research Chemicals Leverandørnr. TRCP7559021MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Progesterone-D9
CAS 15775-74-3
Renhedsgrad noter HPLC
Molekylær formel C21 2H9 H21 O2
Mængde 1 mg
Synonym Pregn-4-ene-3,20-dione-2,2,4,6,6,17,21,21,21-d9, Progesterone-2,2,4,6,6,17,21,21,21-d9 (8CI), 2,2,4,6,6,17,21,21,21-d9-Progesterone
InChI formel InChI=1S/C21H30O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h12,16-19H,4-11H2,1-3H3/t16-,17+,18-,19-,20-,21+/m0/s1/i1D3,4D2,8D2,12D,17D
SMIL [2H]C1=C2C([2H])([2H])C[C@@H]3[C@H](CC[C@@]4(C)[C@H]3CC[C@]4([2H])C(=O)C([2H])([2H])[2H])[C@@]2(C)CC([2H])([2H])C1=O
IUPAC navn (8S,9S,10R,13S,14S,17S)-2,2,4,6,6,17-hexadeuterio-10,13-dimethyl-17-(2,2,2-trideuterioacetyl)-7,8,9,11,12,14,15,16-octahydro-1H-cyclopenta[a]phenanthren-3-one
Molekylvægt (g/mol) 323.52
Formel vægt 323.2811
Procent renhed >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.