missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Progesterone Antibody (6-5E-3F), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-78056R
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Progesterone Monoclonal specifically detects Progesterone in All Species samples. It is validated for ELISA.
Tekniske data
| Progesterone | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| This monoclonal antibody is specific for progesterone. It exhibits minimal cross reactivity with related compounds in ELISA. It reacts with Progesterone-11a-HMS-BSA: 100%; 5-beta-Pregnane-3,20-dione: 48%; 5-alpha-Pregnane-3,20-dione: 26.4%; 17-alpha-Hydroxyprogesterone: 2.5%; 20-alpha-Hydroxyprogesterone: 0.04%. Progesterone is a steroid hormone synthesized from the cholesterol derivative, pregnenolone, in the cortex of the adrenal gland. Progesterone is secreted by the corpus luteum and acts to prepare the endometrium for the implantation of a fertilized egg. During pregnancy, it is secreted by the placenta to prevent spontaneous abortion and to stimulate the development of mammary tissue to produce milk. Thus, progesterone plays a central role in the reproductive events associated with the establishment and maintenance of pregnancy. Luteinized theca cells of normal ovary secrete progesterone. The determination of progesterone concentrations in the body fluids is of great value for endocrinological investigations in women. This monoclonal antibody may prove useful in identification of ovarian tumors. | |
| Store at 4°C in the dark. |
| ELISA | |
| 6-5E-3F | |
| ELISA | |
| 4-Pregnen-3,20-dione | |
| Progesterone hemisuccinate conjugated to bovine serum albumin | |
| 0.1 mL | |
| Primary | |
| All species | |
| IgG1 κ |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion