missing translation for 'onlineSavingsMsg'
Få mere at vide

Profoxydim Lithium Salt, TRC

Artikelnummer. 30010168
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30010168 10 mg
17921375 1 mg
17921275 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30010168 Leverandør Toronto Research Chemicals Leverandørnr. TRCP75589810MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 281664-76-4
Molekylær formel C24 H31 Cl N O4 S . Li
Molekylvægt (g/mol) 471.97
Synonym Profoxydim-lithium, 2-Cyclohexen-1-one, 2-[1-[[2-(4-chlorophenoxy)propoxy]imino]butyl]-3-hydroxy-5-(tetrahydro-2H-thiopyran-3-yl)-, lithium salt (9CI), BAS 625 lithium salt, 2-Cyclohexen-1-one, 2-[1-[[2-(4-chlorophenoxy)propoxy]imino]butyl]-3-hydroxy-5-(tetrahydro-2H-thiopyran-3-yl)-, lithium salt (1:1)
IUPAC navn lithium;2-[(E)-N-[2-(4-chlorophenoxy)propoxy]-C-propylcarbonimidoyl]-3-oxo-5-(thian-3-yl)cyclohexen-1-olate
SMIL [Li+].CCC\C(=N/OCC(C)Oc1ccc(Cl)cc1)\C2=C([O-])CC(CC2=O)C3CCCSC3

Specifications

Kemisk navn eller materiale Profoxydim-lithium
Mængde 10 mg
InChI formel InChI=1S/C24H32ClNO4S.Li/c1-3-5-21(26-29-14-16(2)30-20-9-7-19(25)8-10-20)24-22(27)12-18(13-23(24)28)17-6-4-11-31-15-17;/h7-10,16-18,27H,3-6,11-15H2,1-2H3;/q;+1/p-1/b26-21+;
Formel vægt 471.1822
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.