Learn More
Prodan, TRC
Product Code. 30416136
Change view
Click to view available options
Quantity:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30416136 500 mg
30410050 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30416136 Supplier Toronto Research Chemicals Supplier No. TRCP755865500MG
 Restricted Item
Forventet forsendelse: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 70504-01-7
Molecular Formula C15H17NO
Molecular Weight (g/mol) 227.3
Synonym 1-[6-(Dimethylamino)-2-naphthalenyl]-1-propanone, 2-Propionyl-6-dimethyl-aminonaphthalene, 6-Propionyl-2-(N,N-dimethylamino)naphthalene, 6-Propionyl-2-(dimethylamino)naphthalene, 6-Propanoyl-2-(N,N-dimethylamino)naphthalene, Prodan (fluorophore)
IUPAC Name 1-[6-(dimethylamino)naphthalen-2-yl]propan-1-one
SMILES CCC(=O)c1ccc2cc(ccc2c1)N(C)C
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C15H17NO/c1-4-15(17)13-6-5-12-10-14(16(2)3)8-7-11(12)9-13/h5-10H,4H2,1-3H3
Formula Weight 227.131
Chemical Name or Material Prodan