Få mere at vide
Procaterol Hydrochloride Hydrate, TRC
Artikelnummer. 30138765
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138765 100 mg
17953325 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30138765 Leverandør Toronto Research Chemicals Leverandørnr. TRCP155490100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 81262-93-3
Molekylær formel 2 C16 H22 N2 O3 . 2 Cl H . H2 O
Molekylvægt (g/mol) 671.65
Synonym 2(1H)-Quinolinone, 8-hydroxy-5-[(1R,2S)-1-hydroxy-2-[(1-methylethyl)amino]butyl]-, hydrochloride, hydrate (2:2:1), rel-, 2(1H)-Quinolinone, 8-hydroxy-5-[1-hydroxy-2-[(1-methylethyl)amino]butyl]-, monohydrochloride, hydrate (2:1), (R*,S*)- (9CI), 2(1H)-Quinolinone, 8-hydroxy-5-[1-hydroxy-2-[(1-methylethyl)amino]butyl]-, monohydrochloride, hydrate (2:1), (R*,S*)-(±)-
IUPAC navn 8-hydroxy-5-[(1R,2S)-1-hydroxy-2-(propan-2-ylamino)butyl]-1H-quinolin-2-one;hydrate;dihydrochloride
SMIL O.Cl.Cl.CC[C@H](NC(C)C)[C@H](O)c1ccc(O)c2NC(=O)C=Cc12.CC[C@H](NC(C)C)[C@H](O)c3ccc(O)c4NC(=O)C=Cc34
Tekniske data
Kemisk navn eller materiale Procaterol Hydrochloride Hemihydrate
Mængde 100 mg
InChI formel InChI=1S/2C16H22N2O3.2ClH.H2O/c2*1-4-12(17-9(2)3)16(21)11-5-7-13(19)15-10(11)6-8-14(20)18-15;;;/h2*5-9,12,16-17,19,21H,4H2,1-3H3,(H,18,20);2*1H;1H2/t2*12-,16+;;;/m00.../s1
Formel vægt 670.29