Få mere at vide
Procaine, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Procaine |
| CAS | 59-46-1 |
| Renhedsgrad noter | HPLC |
| Molekylær formel | C13 H20 N2 O2 |
| Mængde | 1 g |
| Synonym | Benzoic acid, 4-amino-, 2-(diethylamino)ethyl ester, Benzoic acid, p-amino-, 2-(diethylamino)ethyl ester (8CI), 2-(Diethylamino)ethyl p-aminobenzoate, 2-Diethylaminoethyl 4-aminobenzoate, 4-Aminobenzoic acid 2-(diethylamino)ethyl ester, 4-Aminobenzoic acid diethylaminoethyl ester, Diethylaminoethyl p-aminobenzoate, Duracaine, NSC 169497, Nissocaine, Pfizerpen AS, Procain, Procaine, Procaine base, SP 01, Spinocaine, Vitamin H3, p-Aminobenzoic acid 2-diethylaminoethyl ester, β-(Diethylamino)ethyl p-aminobenzoate, β-Diethylaminoethyl 4-aminobenzoate |
| InChI formel | InChI=1S/C13H20N2O2/c1-3-15(4-2)9-10-17-13(16)11-5-7-12(14)8-6-11/h5-8H,3-4,9-10,14H2,1-2H3 |
| SMIL | CCN(CC)CCOC(=O)c1ccc(N)cc1 |
| IUPAC navn | 2-(diethylamino)ethyl 4-aminobenzoate |
| Molekylvægt (g/mol) | 236.31 |
| Vis mere |