missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Procaine Penicillin G Hydrate, TRC
Specifications
Specifications
| Renhed | >95% (HPLC) |
| Format | Neat |
| Molekylær formel | C29H40N4O7S |
| Molekylvægt (g/mol) | 588.72 |
| InChI nøgle | KZDCMKVLEYCGQX-UDPGNSCCSA-N |
| SMIL | O.CCN(CC)CCOC(=O)C1=CC=C(N)C=C1.CC1(C)S[C@@H]2[C@H](NC(=O)CC3=CC=CC=C3)C(=O)N2[C@H]1C(O)=O |
| IUPAC navn | (2S,5R,6R)-3,3-dimethyl-7-oxo-6-(2-phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid 2-(diethylamino)ethyl 4-aminobenzoate hydrate |
| Produkttype | API |
| Mængde | 5 g |
| Forsendelsestilstand | Room Temperature |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?