Få mere at vide
Pro-Gln-OH, TRC
Artikelnummer. 30425867
Skift visning
Mængde:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30425867 50 mg
30427847 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30425867 Leverandør Toronto Research Chemicals Leverandørnr. TRCP83999850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Pro-Gln-OH
CAS 18668-08-1
Molekylær formel C10 H17 N3 O4
Mængde 50 mg
InChI formel InChI=1S/C10H17N3O4/c11-8(14)4-3-7(10(16)17)13-9(15)6-2-1-5-12-6/h6-7,12H,1-5H2,(H2,11,14)(H,13,15)(H,16,17)/t6-,7-/m0/s1
SMIL NC(=O)CC[C@H](NC(=O)[C@@H]1CCCN1)C(=O)O
IUPAC navn (2S)-5-amino-5-oxo-2-[[(2S)-pyrrolidine-2-carbonyl]amino]pentanoic acid
Molekylvægt (g/mol) 243.26
Formel vægt 243.122