missing translation for 'onlineSavingsMsg'
Learn More

Chemsplash Pro+4 Type 456 Coverall with Feet, Taped
GREENER_CHOICE

Pro +4 Disposable Coverall with Feet is made from a 67GSM microporous laminated Cat III Type 4B/5B/6B fabric.

1940.00 DKK

Specifications

Product Type Cleanroom Coveralls
Gender Unisex
Material Microporous Composite Fabric
Color White
Disposable Single-use
View More Specs

Products 6
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
30058187
GREENER_CHOICE
View Documents
Chemsplash
2599.400.004
Small
1940.00 DKK
Pack of 25
Only null left
Estimated Shipment: 01-08-2025
Add to basket
 
30058418
GREENER_CHOICE
View Documents
Chemsplash
2599.400.005
Medium
1940.00 DKK
Pack of 25
Only null left
Estimated Shipment: 01-08-2025
Add to basket
 
30058457
GREENER_CHOICE
View Documents
Chemsplash
2599.400.006
Large
1940.00 DKK
Pack of 25
Only null left
Estimated Shipment: 01-08-2025
Add to basket
 
30058134
GREENER_CHOICE
View Documents
Chemsplash
2599.400.007
X-Large
1940.00 DKK
Pack of 25
Only null left
Estimated Shipment: 01-08-2025
Add to basket
 
30058316
GREENER_CHOICE
View Documents
Chemsplash
2599.400.008
2X-Large
1940.00 DKK
Pack of 25
Only null left
Estimated Shipment: 01-08-2025
Add to basket
 
30058140
GREENER_CHOICE
View Documents
Chemsplash
2599.400.009
3X-Large
1940.00 DKK
Pack of 25
Only null left
Estimated Shipment: 01-08-2025
Add to basket
 
Description

Description

Chemsplash Pro +4 Disposable Coverall with Feet

  • 67GSM Microporous Laminate Fabric
  • Coverall Comes with Feet Attached
  • Fully Taped Seams
  • Three Piece Hood
  • Elasticated Cuffs with Thumb Loops
  • Elasticated Hood, Back and Ankles
  • Two Way Zip with Zip Flap
  • Silicone and Latex Free, PFAS Free
  • Low Linting, Anti-Static
  • Conforms to ISO 14644-1:2015 Cleanroom 5 - 9

Description: Chemsplash Pro +4 Disposable Coverall with Feet is made from a 67GSM microporous laminated Cat III Type 4B/5B/6B fabric. All seams are taped offering a stronger and more effective barrier to liquids and particulate matter. Chemsplash Pro +4 fabric is both anti-static to EN1149-5:2008 and is resistant to infectious agents under EN14126, complying with resistance to viral and bacterial infection. Making it an ideal choice for Pharmaceutical, Cleanroom, Semi-Conductor manufacturing and Infection Contamination Control. Conforms to ISO 14644-1:2015 Cleanroom 5 - 9.

Specifications

Specifications

Cleanroom Coveralls
Microporous Composite Fabric
Single-use
Attached Boots
Hooded
Ties
Non-sterile
Yes
Class 100-100,000 (ISO 5-8)
Unisex
White
Elastic
Elastic, Thumb Loop
3-panel
Stitched, Taped
Liquid
None
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.