missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRKDC(phospho T2609) & PRKDC Protein Phosphorylation Antibody Pair, Abnova™
Antibody Pair
Brand: Abnova DP0191.1Set
This item is not returnable.
View return policy
Description
This protein phosphorylation antibody pair set comes with two antibodies, one against the PRKDC protein, and the other against the specific T2609 phosphorylated site of PRKDC for use in in situ Proximity Ligation Assay.Specifications
PRKDC | |
Unconjugated | |
DNA-PKcs/DNAPK/DNPK1/HYRC/HYRC1/XRCC7/p350 | |
1 Set | |
Primary | |
Antibody pair set content: 1. Phospho-PRKDC T2609 rabbit polyclonal antibody (20 ul) In PBS (without Mg2+ and Ca2+), 150 mM NaCl, pH 7.4 (0.02% sodium azide, 50% glycerol) 2. PRKDC mouse monoclonal antibody (40 ug) In 1x PBS, pH 7.2 *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
In situ PLA | |
protein kinase, DNA-activated, catalytic polypeptide | |
PRKDC | |
RUO | |
5591 | |
Human | |
Ab Pair |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PRKDC(phospho T2609) & PRKDC Protein Phosphorylation Antibody Pair, Abnova™ > 1 Set; Unconjugated
Spot an opportunity for improvement?Share a Content Correction