missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ Human PRKCG (PKC gamma), GST Tag Recombinant Protein

Artikelnummer. 17727517
Klik for at se tilgængelige muligheder
Mængde:
1 mg
10 μg
100 μg
Denne vare kan ikke returneres. Se returpolicy

Artikelnummer. 17727517

Brand: Invitrogen™ A41918

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length, Active The gene accession number for PRKCG (PKC gamma) is NP_002730.1.

The PKC family of serine/threonine kinases, including PRKCG (PKC gamma), is activated intracellularly by signal transduction pathways. In humans, at least 12 different PKC polypeptides have been identified. These isoforms differ in primary structure, tissue distribution, subcellular localization, mode of action in vitro, response to extracellular signals, and substrate specificity. PKC alpha, beta I, beta II, and gamma form the conventional family; their activities are Ca2+- and phospholipid-dependent. Protein kinase C can be activated by calcium and second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play distinct roles in cells. PKC gamma is one of the PKC family members. This protein kinase is expressed solely in the brain and spinal cord and its localization is restricted to neurons. It has been demonstrated that several neuronal functions, including long term potentiation (LTP) and long term depression (LTD), specifically require this kinase. Knockout studies in mice also suggest that this kinase may be involved in neuropathic pain development. Defects in this protein have been associated with neurodegenerative disorder spinocerebellar ataxia-14 (SCA14).
TRUSTED_SUSTAINABILITY

Tekniske data

Adgangsnummer P05129
Koncentration See Label
Til brug med (applikation) Kinase Assay
Formulering 50mM tris HCl with 20% glycerol, 100mM NaCl, 15mM glutathione, 5mM DTT and no preservative; pH 8
Gen-id (Entrez) 5582
Molekylvægt (g/mol) 112.3 kDa
Navn Human PRKCG (PKC gamma), GST Tag
pH-område 8
Oprensningsmetode Purified
Mængde 100 μg
Opbevaringskrav -80°C, Avoid Freeze/Thaw Cycles
Regulatorisk status RUO
Gene Alias PKC; Pkcc; PKCG; PKCgamma; PKC-gamma; PKCI; Prkc; Prkcc; PRKCG; protein kinase C gamma; protein kinase C gamma type; protein kinase C type I (gamma type); protein kinase C, gamma; RATPKCI; SCA14
Fælles navn PKC gamma
Gen symbol PRKCG
KinaseFamily PKC Kinase Family
Produkttype Protein
Konjugeret Unconjugated
Arter Human
Rekombinant Recombinant
Protein tag GST-tag
Sekvens Full length
Indhold og opbevaring -80°C, Avoid Freeze/Thaw Cycles
Forsendelsestilstand Dry Ice
Udtrykssystem Insect cells
Protein form Full Length, Active
Form Liquid
Protein undertype Serine⁄Threonine Kinases
Forskningskategori Drug Development
Vis mere Vis mindre
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.