missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PRKCB2 (PKC beta II) Recombinant Human Protein

The PKC family of serine/threonine kinases, including PRKCB2 (PKC beta II), is activated intracellularly by signal transduction pathways.

Brand:  Invitrogen™ PR1461C

Additional Details : Weight : 0.01000kg

Product Code. 15408373

  • 9370.00 DKK / 100µg


Estimated Shipment date
from Supplier 23-07-2018
Add to basket

Description

Description

In humans, at least 12 different PKC polypeptides have been identified. These isoforms differ in primary structure, tissue distribution, subcellular localization, mode of action in vitro, response to extracellular signals, and substrate specificity. PKC alpha, beta I, beta II and gamma form the conventional family; their activities are Ca2+- and phospholipid-dependent.

Specifications

Specifications

NP_002729
Frozen
77kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
PKC Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50% glycerol, 20mM HEPES, pH 7.5, 2mM EDTA, 2mM EGTA, 5mM DTT, 250mM NaCl, 0.05% Triton X-100
100μg
RUO - research use only
PRKCB2 (PKC beta II)
Baculovirus
Human
None
Videos