Learn More
Epredia™ PrintMate™ Slotted Cassettes in Pre-packed Tubes, Aqua
Epredia™ PrintMate Cassettes™ are designed to contain incised tissue safely and securely for tissue processing, embedding and sectioning. Packed in tubes, PrintMate Cassettes are intended for high throughput printing, and provide exceptional scanning accuracy and reliability when used with the Epredia™ PrintMate™ and Epredia™ SlideMate™ barcode scanner.
Brand: Epredia™ A84210033
Additional Details : Weight : 4.50000kg
Specifications
Slotted Pore | |
Yes | |
Gray | |
Biopsy Cassette |