Learn More
Epredia™ PrintMate™ Slottede kassetter
Shop All Epredia Products
Click to view available options
Farve:
Aqua
Beige
Blå
Blå
Grå
Grøn
Grå
Grøn
Gul
Hvid
Lavendel
Lilla
Lysegrøn
Lyserød
Orange
Pink (Pink)
Tan
Unit Size:
Pakke med 1000
Pakke med 750
Mængde:
10 x 75 kassetter (750)/cs
4 x 250 kassetter (1000)/cs
Description
Designed for performance and user convenience, PrintMate Slotted Cassettes have been optimized for superior workflow and printing with the PrintMate Classic and PrintMate AS printing systems. The even, textured printing face provides the optimal surface for high-quality printing every time. For additional information, the right and left sides are textured for clear handwriting with suitable pens. The lid catches have been adjusted for ease of use with low opening and closing force, while still giving the security needed while processing.
Available in tubes for automatic printing or loose for manual use, and in twelve colors for enhanced workflow identification.
Available in tubes for automatic printing or loose for manual use, and in twelve colors for enhanced workflow identification.
Specifications
Specifications
| Pore stil | Slotted Pore |
| PrintMate kompatibel | Yes |
| Farve | Lilla |
| Produkttype | Biopsy Cassette |