Få mere at vide
Primidone-d5, TRC
Artikelnummer. 17977359
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17977359 10 mg
17959079 5 mg
17995115 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 17977359 Leverandør Toronto Research Chemicals Leverandørnr. TRCP73400210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 73738-06-4
Molekylær formel C12 2H5 H9 N2 O2
Molekylvægt (g/mol) 223.28
Synonym 4,6(1H,5H)-Pyrimidinedione, 5-(ethyl-d5)dihydro-5-phenyl- (9CI), Primidone-d5, Primidone-D5 (ethyl-D5), 5-(Ethyl-d5)-5-phenyldihydropyrimidine-4,6(1H,5H)-dione, 2-Desoxyphenobarbital-d5
IUPAC navn 5-(1,1,2,2,2-pentadeuterioethyl)-5-phenyl-1,3-diazinane-4,6-dione
SMIL [2H]C([2H])([2H])C([2H])([2H])C1(C(=O)NCNC1=O)c2ccccc2
Tekniske data
Kemisk navn eller materiale Primidone-d5
Renhedsgrad noter HPLC
Mængde 10 mg
InChI formel InChI=1S/C12H14N2O2/c1-2-12(9-6-4-3-5-7-9)10(15)13-8-14-11(12)16/h3-7H,2,8H2,1H3,(H,13,15)(H,14,16)/i1D3,2D2
Formel vægt 223.1369
Procent renhed >95%