Få mere at vide
Prenylamine Lactate, Mikromol™
Beskrivelse
Prenylamine Lactate
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Prenylamine Lactate |
| CAS | 69-43-2 |
| Analyt- eller komponentnavne | Prenylamine Lactate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C24 H27 N . C3 H6 O3 |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Agozol, Angormin, Bismetin, Carditin-Same, Coredamin, Corontin, Crepasin, Daxauten, Difril, Hostaginan, Incoran, Irrorin, Lactamin, Plactamin, Prenylamine lactate, Reocorin, Roinin, Seccidin, Sedolaton, Synadrin, dl-Prenylamine lactate, Propanoic acid, 2-hydroxy-, compd. with N-(1-methyl-2-phenylethyl)-gamma-phenylbenzenepropanamine (1:1), Lactic acid, compd. with N-(3,3-diphenylpropyl)-alpha-methylphenethylamine (1:1) (8CI), Lactic acid, compd. with N-(3,3-diphenylpropyl)-alpha-methylphenethylamine (7CI), Lactic acid, salt with N-(3,3-diphenylpropyl)-alpha-methylphenethylamine (6CI), Benzenepropanamine, N-(1-methyl-2-phenylethyl)-gamma-phenyl-, 2-hydroxypropanoate (9CI), Phenethylamine, N-(3,3-diphenylpropyl)-alpha-methyl-, lactate (8CI), N-(3,3-Diphenylpropyl)-alpha-methylphenethylamine lactate |
| InChI formel | InChI=1S/C24H27N.C3H6O3/c1-20(19-21-11-5-2-6-12-21)25-18-17-24(22-13-7-3-8-14-22)23-15-9-4-10-16-23;1-2(4)3(5)6/h2-16,20,24-25H,17-19H2,1H3;2,4H,1H3,(H,5,6) |
| SMIL | CC(Cc1ccccc1)NCCC(c2ccccc2)c3ccccc3.CC(O)C(=O)O |
| IUPAC navn | 3,3-diphenyl-N-(1-phenylpropan-2-yl)propan-1-amine;2-hydroxypropanoic acid |
| Vis mere |