Få mere at vide
Pregnenolone-d4, TRC
Artikelnummer. 30412292
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30412292 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30412292 Leverandør Toronto Research Chemicals Leverandørnr. TRCP71220210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Pregnenolone-d4
CAS 61574-54-7
Renhedsgrad noter HPLC
Molekylær formel C21 2H4 H28 O2
Mængde 10 mg
Synonym Pregn-5-en-20-one-17,21,21,21-d4','3-hydroxy-','(3beta)-, Pregnenolone-D4
InChI formel InChI=1S/C21H32O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h4,15-19,23H,5-12H2,1-3H3/t15-,16-,17+,18-,19-,20-,21+/m0/s1/i1D3,17D
SMIL [2H]C([2H])([2H])C(=O)[C@@]1([2H])CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC navn 2,2,2-trideuterio-1-[(3S,8S,9S,10R,13S,14S,17S)-17-deuterio-3-hydroxy-10,13-dimethyl-1,2,3,4,7,8,9,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-17-yl]ethanone
Molekylvægt (g/mol) 320.5
Formel vægt 320.2653
Procent renhed >95