missing translation for 'onlineSavingsMsg'
Få mere at vide

Pregnenolone-16-ene Acetate Oxime, TRC

Artikelnummer. 30421857
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30421857 1 g
30418145 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30421857 Leverandør Toronto Research Chemicals Leverandørnr. TRCP7122301G

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 2174-13-2
Molekylær formel C23 H33 N O3
Molekylvægt (g/mol) 371.51
Synonym 16-Dehydropregnenolone acetate oxime, Pregna-5,16-dien-20-one, 3-(acetyloxy)-, 20-oxime, (3β)-, Pregna-5,16-dien-20-one, 3β-hydroxy-, acetate, oxime (6CI), Pregna-5,16-dien-20-one, 3β-hydroxy-, oxime, acetate (ester) (8CI), (3β)-3-(Acetyloxy)pregna-5,16-dien-20-one 20-oxime, NSC 224249
IUPAC navn [(3S,8R,9S,10R,13S,14S)-17-[(Z)-N-hydroxy-C-methylcarbonimidoyl]-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15-decahydro-1H-cyclopenta[a]phenanthren-3-yl] acetate
SMIL CC(=O)O[C@H]1CC[C@]2(C)[C@H]3CC[C@@]4(C)[C@@H](CC=C4\C(=N/O)\C)[C@@H]3CC=C2C1

Tekniske data

Kemisk navn eller materiale 16-Dehydropregnenolone Acetate Oxime
Mængde 1 g
InChI formel InChI=1S/C23H33NO3/c1-14(24-26)19-7-8-20-18-6-5-16-13-17(27-15(2)25)9-11-22(16,3)21(18)10-12-23(19,20)4/h5,7,17-18,20-21,26H,6,8-13H2,1-4H3/b24-14-/t17-,18-,20-,21-,22-,23+/m0/s1
Formel vægt 371.246
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.