Få mere at vide
Pregnanediol (5beta-Pregnane-3alpha,20(S)-diol), Mikromol™
Artikelnummer. 30050745
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050745 25 mg
17989544 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30050745 Leverandør Mikromol Leverandørnr. MM0222.140025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Pregnanediol (5beta-Pregnane-3alpha,20(S)-diol)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Pregnanediol
CAS 80-92-2
Analyt- eller komponentnavne Pregnanediol
Forsendelsestilstand Room Temperature
Molekylær formel C21 H36 O2
Certificeringer/compliance ISO 17025
Synonym Pregnanediol, 5beta-Pregnane-3alpha,20(S)-diol, 5beta-Pregnane-3alpha,20alph-diol, 3alpha,20alpha-Dihydroxy-5beta-pregnane, NSC 1612, NSC 47462
InChI formel InChI=1S/C21H36O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h13-19,22-23H,4-12H2,1-3H3/t13-,14+,15+,16-,17+,18-,19-,20-,21+/m0/s1
SMIL C[C@H](O)[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC navn (3R,5R,8R,9S,10S,13S,14S,17S)-17-[(1S)-1-hydroxyethyl]-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-ol
Molekylvægt (g/mol) 320.51
Mængde 25 mg
Formel vægt 320.2715 g/mol