Få mere at vide
Pregna-4,9(11)-diene-3,20-dione (90%), TRC
Artikelnummer. 30410905
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30410905 100 mg
30427556 50 mg
30427275 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30410905 Leverandør Toronto Research Chemicals Leverandørnr. TRCP704970100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 17652-16-3
Molekylær formel C21 H28 O2
Molekylvægt (g/mol) 312.45
Synonym Pregna-4,9(11)-diene-3,20-dione, 9-Dehydroprogesterone, δ9(11)-Progesterone, Progesterone Imp. K (EP)
IUPAC navn (8S,10S,13S,14S,17S)-17-acetyl-10,13-dimethyl-1,2,6,7,8,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-3-one
SMIL CC(=O)[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)C3=CC[C@]12C
Tekniske data
Kemisk navn eller materiale Pregna-4,9(11)-diene-3,20-dione
Mængde 100 mg
InChI formel InChI=1S/C21H28O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h9,12,16-18H,4-8,10-11H2,1-3H3/t16-,17+,18-,20-,21+/m0/s1
Formel vægt 312.2089